accessibility menu, dialog, popup

UserName

H3CCH3OOHCH3CH3HOCH3CH3HOFe

CAS RN 331720-17-3

IUPAC Name: 2-ethylhexanoic acid bis(propan-2-ol) iron
Synonyms: 2-ethylhexanoic acid bis(isopropyl alcohol) iron
Molecular Weight (g/mol): 320.25
Molecular Formula: C14H32FeO4
InChi Key: GCDBQBNMPOKFEM-UHFFFAOYNA-N
SMILES: [Fe].CC(C)O.CC(C)O.CCCCC(CC)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results