CAS RN 33136-66-2
CAS RN 33136-66-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-1-Ferrocenylethanol, 99%, Thermo Scientific™
CAS: 33136-66-2 Molecular Formula: C12H14FeO Molecular Weight (g/mol): 230.09 MDL Number: MFCD01074672 IUPAC Name: (S)-1-Ferrocenylethanol SMILES: [Fe].c1cccc1.C[C@H](O)c1cccc1