accessibility menu, dialog, popup

UserName

H3CCH3OHH3C

CAS RN 3228-02-2

IUPAC Name: 3-methyl-4-(propan-2-yl)phenol
Synonyms: o-cymen-5-ol p-thymol frecide o-cymen-5y-ol biosal (antibacterial) isopropyl cresols
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: IJALWSVNUBBQRA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results