accessibility menu, dialog, popup

UserName

CAS RN 321531-51-5

CH3NOClNOO

CAS RN 321531-51-5

IUPAC Name: N-benzyl-4-chloro-N-ethyl-3-nitrobenzamide
Synonyms: N-benzyl-4-chloro-N-ethyl-3-nitrobenzamide
Molecular Weight (g/mol): 318.76
Molecular Formula: C16H15ClN2O3
InChi Key: VDTSYXSLPINNCH-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results