accessibility menu, dialog, popup

UserName

CAS RN 31805-83-1

H3CSOHSCH3

CAS RN 31805-83-1

IUPAC Name: 1,3-bis(methylsulfanyl)propan-2-ol
Synonyms: 1,3-bis(methylsulfanyl)propan-2-ol
Molecular Weight (g/mol): 152.27
Molecular Formula: C5H12OS2
InChi Key: BTQNSLFPJKRBQK-UHFFFAOYSA-N
SMILES: CSCC(O)CSC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results