CAS RN 31582-45-3
CAS RN 31582-45-3
IUPAC Name:
2,2,6,6-tetramethylpiperidin-4-yl 2-methylprop-2-enoate
Synonyms:
2,2,6,6-tetramethylpiperidin-4-yl 2-methylprop-2-enoate
Molecular Weight (g/mol):
225.33
Molecular Formula:
C13H23NO2
InChi Key:
UFLXKQBCEYNCDU-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OC1CC(C)(C)NC(C)(C)C1
1
–
3
of
3
results
1
2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.