missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TRC

Product Code. 30456354
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456354 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456354 Supplier Toronto Research Chemicals Supplier No. TRCT30464025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidyl Methacrylate
CAS 31582-45-3
Molecular Formula C13H23NO2
Quantity 25 g
Synonym 2-Methyl-2-propenoic Acid 2,2,6,6-Tetramethyl-4-piperidinyl Ester, Methacrylic Acid 2,2,6,6-Tetramethyl-4-piperidyl Ester, 2,2,6,6-Tetramethyl-4-piperidinyl 2-methyl-2-propenoate, 4-(Methacryloyloxy)-2,2,6,6-tetramethylpiperidine, ADK Stab LA 87, Fancryl FA 712HM, Mark LA 87
InChI Formula InChI=1S/C13H23NO2/c1-9(2)11(15)16-10-7-12(3,4)14-13(5,6)8-10/h10,14H,1,7-8H2,2-6H3
SMILES CC(=C)C(=O)OC1CC(C)(C)NC(C)(C)C1
IUPAC Name (2,2,6,6-tetramethylpiperidin-4-yl) 2-methylprop-2-enoate
Molecular Weight (g/mol) 225.33
Formula Weight 225.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.