missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2,6,6-Tetramethyl-4-piperidyl Methacrylate |
| CAS | 31582-45-3 |
| Molecular Formula | C13H23NO2 |
| Quantity | 5 g |
| Synonym | 2-Methyl-2-propenoic Acid 2,2,6,6-Tetramethyl-4-piperidinyl Ester, Methacrylic Acid 2,2,6,6-Tetramethyl-4-piperidyl Ester, 2,2,6,6-Tetramethyl-4-piperidinyl 2-methyl-2-propenoate, 4-(Methacryloyloxy)-2,2,6,6-tetramethylpiperidine, ADK Stab LA 87, Fancryl FA 712HM, Mark LA 87 |
| InChI Formula | InChI=1S/C13H23NO2/c1-9(2)11(15)16-10-7-12(3,4)14-13(5,6)8-10/h10,14H,1,7-8H2,2-6H3 |
| SMILES | CC(=C)C(=O)OC1CC(C)(C)NC(C)(C)C1 |
| IUPAC Name | (2,2,6,6-tetramethylpiperidin-4-yl) 2-methylprop-2-enoate |
| Molecular Weight (g/mol) | 225.33 |
| Formula Weight | 225.1729 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?