CAS RN 312959-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, CTC-Q-PHOS, 95%, Thermo Scientific Chemicals
CAS: 312959-24-3 Molecular Formula: C48H47FeP Molecular Weight (g/mol): 710.72 MDL Number: MFCD03788940 IUPAC Name: 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene SMILES: [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1