CAS RN 305350-87-2
CAS RN 305350-87-2
IUPAC Name:
3-amino-3-[(4-aminophenyl)sulfanyl]-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile
Synonyms:
3-amino-3-[(4-aminophenyl)sulfanyl]-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile
Molecular Weight (g/mol):
335.35
Molecular Formula:
C16H12F3N3S
InChi Key:
JLOXTZFYJNCPIS-UHFFFAOYSA-N
SMILES:
NC(SC1=CC=C(N)C=C1)=C(C#N)C1=CC=CC=C1C(F)(F)F