accessibility menu, dialog, popup

UserName

H3COCH3OCH3OSb

CAS RN 29671-18-9

IUPAC Name: antimony(3+) trimethanolate
Synonyms: antimony(3+) tris(methoxide)
Molecular Weight (g/mol): 214.86
Molecular Formula: C3H9O3Sb
InChi Key: KEUYHGXCOWNTEJ-UHFFFAOYSA-N
SMILES: [Sb+3].C[O-].C[O-].C[O-]
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)