accessibility menu, dialog, popup

UserName

CH3NHSOOFCH3

CAS RN 288154-65-4

IUPAC Name: N-(3,4-dimethylphenyl)-4-fluorobenzene-1-sulfonamide
Synonyms: N-(3,4-dimethylphenyl)-4-fluorobenzenesulfonamide
Molecular Weight (g/mol): 279.33
Molecular Formula: C14H14FNO2S
InChi Key: IJCSKFYHYFBPNB-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(NS(=O)(=O)C2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results