CAS RN 28739-42-6
IUPAC Name:
3-(5-methyl-1H-pyrazol-4-yl)propan-1-amine
Synonyms:
3-(3-methyl-2H-pyrazol-4-yl)propan-1-amine
Molecular Weight (g/mol):
139.2
Molecular Formula:
C7H13N3
InChi Key:
BYGCQGLHPCERCH-UHFFFAOYSA-N
SMILES:
CC1=C(CCCN)C=NN1
1
–
1
of
1
results
1
3-(5-Methyl-1H-pyrazol-4-yl)propylamine, Technical Grade, Thermo Scientific™
CAS: 28739-42-6 Molecular Formula: C7H13N3 Molecular Weight (g/mol): 139.202 InChI Key: BYGCQGLHPCERCH-UHFFFAOYSA-N Synonym: 3-5-methyl-1h-pyrazol-4-yl propan-1-amine,3-5-methyl-1h-pyrazol-4-yl propylamine,3-3-methyl-2h-pyrazol-4-yl propan-1-amine,4-3-aminopropyl-5-methylpyrazole,3-5-methylpyrazol-4-yl propylamine,1h-pyrazole-4-propanamine,3-methyl,4-3-aminopropyl-5-methyl-1h-pyrazole, tech PubChem CID: 2736943 IUPAC Name: 3-(5-methyl-1H-pyrazol-4-yl)propan-1-amine SMILES: CC1=C(C=NN1)CCCN