accessibility menu, dialog, popup

UserName

ONNN

CAS RN 27996-86-7

IUPAC Name: 4-(1H-1,2,4-triazol-1-yl)benzaldehyde
Synonyms: 4-(1,2,4-triazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 173.18
Molecular Formula: C9H7N3O
InChi Key: TVEJNWMWDIXPAX-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results