accessibility menu, dialog, popup

UserName

H3COHOFFF

CAS RN 276861-64-4

IUPAC Name: [3-methoxy-4-(trifluoromethyl)phenyl]methanol
Synonyms: [3-methoxy-4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: KGRHSPHJFOSYLA-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(CO)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results