accessibility menu, dialog, popup

UserName

OHOO

CAS RN 2745-26-8

IUPAC Name: 2-(furan-2-yl)acetic acid
Synonyms: furan-2-yl-acetic acid
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: VYSRZETUSAOIMP-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)