CAS RN 2726492-72-2
CAS RN 2726492-72-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Buspirone EP Impurity J, TRC
CAS: 2726492-72-2 Molecular Formula: C34H52N6O5 Molecular Weight (g/mol): 624.81 Synonym: 4-(7,9-Dioxo-8-azaspiro[4.5]decan-8-yl)butyl 2-(1-(2-Oxo-2-((4-(4-(pyrimidin-2-yl)piperazin-1-yl)butyl)amino)ethyl)cyclopentyl)acetate SMILES: O=C(CC1(CC(OCCCCN(C(CC2(CCCC2)C3)=O)C3=O)=O)CCCC1)NCCCCN(CC4)CCN4C5=NC=CC=N5