accessibility menu, dialog, popup

UserName

CAS RN 26893-73-2

CH3ONOHO

CAS RN 26893-73-2

IUPAC Name: 6-methoxypyridine-2-carboxylic acid
Synonyms: 6-methoxypyridine-2-carboxylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: KSWBODXXZITTPO-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results