accessibility menu, dialog, popup

UserName

CAS RN 267225-27-4

OOHH2NBr

CAS RN 267225-27-4

IUPAC Name: 2-amino-3-(2-bromophenyl)propanoic acid
Synonyms: 2-amino-3-(2-bromophenyl)propanoic acid
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: JFVLNTLXEZDFHW-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1Br)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)