accessibility menu, dialog, popup

UserName

CAS RN 26214-68-6

IUPAC Name: 1-methyl-1H-pyrrole-2-carbonyl chloride
Synonyms: 1-methylpyrrole-2-carbonyl chloride
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: JOYDZQJINHJNPM-UHFFFAOYSA-N
SMILES: CN1C=CC=C1C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results