accessibility menu, dialog, popup

UserName

CAS RN 261763-20-6

OHClFFF

CAS RN 261763-20-6

IUPAC Name: [2-chloro-3-(trifluoromethyl)phenyl]methanol
Synonyms: [2-chloro-3-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 210.58
Molecular Formula: C8H6ClF3O
InChi Key: MQUXXVLMXCHGDZ-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results