CAS RN 261762-97-4
CAS RN 261762-97-4
IUPAC Name:
2-(5-chloro-2-fluorophenyl)acetic acid
Synonyms:
(5-chloro-2-fluorophenyl)acetic acid
Molecular Weight (g/mol):
188.58
Molecular Formula:
C8H6ClFO2
InChi Key:
NOKLOLLCDCEFAK-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=C(F)C=CC(Cl)=C1
1
–
1
of
1
results
1
5-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™
CAS: 261762-97-4 Molecular Formula: C8H6ClFO2 Molecular Weight (g/mol): 188.582 MDL Number: MFCD01631439 InChI Key: NOKLOLLCDCEFAK-UHFFFAOYSA-N Synonym: 5-chloro-2-fluorophenylacetic acid,2-5-chloro-2-fluorophenyl acetic acid,5-chloro-2-fluorophenyl acetic acid,pubchem7141,5-chloro-2-fluoro-phenylacetic acid,2-5-chloro-2-fluorophenyl aceticacid,benzeneacetic acid,5-chloro-2-fluoro PubChem CID: 2773723 IUPAC Name: 2-(5-chloro-2-fluorophenyl)acetic acid SMILES: C1=CC(=C(C=C1Cl)CC(=O)O)F