accessibility menu, dialog, popup

UserName

CAS RN 2585-24-2

H3CNHONOO

CAS RN 2585-24-2

IUPAC Name: 4-nitro-N-propylbenzamide
Synonyms: 4-nitro-N-propylbenzamide
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: MDRVSZJDSUDYGP-UHFFFAOYSA-N
SMILES: CCCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results