accessibility menu, dialog, popup

UserName

CAS RN 25790-35-6

H3COOO

CAS RN 25790-35-6

IUPAC Name: 1-(furan-2-yl)butane-1,3-dione
Synonyms: 1-(furan-2-yl)butane-1,3-dione
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: GPYKJDYMMUIUFG-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)C1=CC=CO1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)