accessibility menu, dialog, popup

UserName

CAS RN 256507-00-3

OSONHN

CAS RN 256507-00-3

IUPAC Name: 5-(furan-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5-(furan-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 218.23
Molecular Formula: C10H6N2O2S
InChi Key: JTIBBSSPYDCKBN-UHFFFAOYSA-N
SMILES: O=C1NC=NC2=C1C(=CS2)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results