accessibility menu, dialog, popup

UserName

PdPPClClPhPhPhPh

CAS RN 253157-79-8

IUPAC Name: palladium(2+) [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane dichloride
Synonyms: palladium(2+) binap dichloride
Molecular Weight (g/mol): 800.01
Molecular Formula: C44H32Cl2P2Pd
InChi Key: VDHAUMFISVWIRX-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results