accessibility menu, dialog, popup

UserName

NN

CAS RN 253-82-7

IUPAC Name: quinazoline
Synonyms: quinazoline benzo(a)pyrimidine phenmiazine 5,6-benzopyrimidine chinazolin benzo[a]pyrimidine
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: JWVCLYRUEFBMGU-UHFFFAOYSA-N
SMILES: C1=CC=C2N=CN=CC2=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)