CAS RN 241799-10-0
CAS RN 241799-10-0
IUPAC Name:
5-cyclopropyl-N-(diaminomethylidene)-1-(quinolin-5-yl)-1H-pyrazole-4-carboxamide dihydrochloride
Synonyms:
5-cyclopropyl-N-(diaminomethylidene)-1-(quinolin-5-yl)pyrazole-4-carboxamide dihydrochloride
Molecular Weight (g/mol):
393.27
Molecular Formula:
C17H18Cl2N6O
InChi Key:
ARZNPJAGKWHEHT-UHFFFAOYSA-N
SMILES:
Cl.Cl.NC(N)=NC(=O)C1=C(C2CC2)N(N=C1)C1=C2C=CC=NC2=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Zoniporide dihydrochloride
CAS: 241799-10-0 Molecular Formula: C17H18Cl2N6O