accessibility menu, dialog, popup

UserName

CAS RN 235088-67-2

H2NFFF

CAS RN 235088-67-2

IUPAC Name: 1-(2,3,4-trifluorophenyl)methanamine
Synonyms: 1-(2,3,4-trifluorophenyl)methanamine
Molecular Weight (g/mol): 161.13
Molecular Formula: C7H6F3N
InChi Key: YIWUAPISITUDBY-UHFFFAOYSA-N
SMILES: NCC1=CC=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results