accessibility menu, dialog, popup

UserName

H3COONOO

CAS RN 23136-39-2

IUPAC Name: 1-(5-nitro-1-benzofuran-2-yl)ethan-1-one
Synonyms: 1-(5-nitro-1-benzofuran-2-yl)ethanone
Molecular Weight (g/mol): 205.17
Molecular Formula: C10H7NO4
InChi Key: RMXFQPLULZFJMH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC(=CC=C2O1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results