accessibility menu, dialog, popup

UserName

CAS RN 230295-16-6

HOFFFF

CAS RN 230295-16-6

IUPAC Name: [3-fluoro-4-(trifluoromethyl)phenyl]methanol
Synonyms: [3-fluoro-4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: MTBRNORZNYPTIU-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results