CAS RN 230-07-9
CAS RN 230-07-9
IUPAC Name:
4,7-phenanthroline
Synonyms:
4,7-phenanthroline
Molecular Weight (g/mol):
180.21
Molecular Formula:
C12H8N2
InChi Key:
DATYUTWESAKQQM-UHFFFAOYSA-N
SMILES:
C1=CN=C2C=CC3=NC=CC=C3C2=C1