CAS RN 2292-16-2
CAS RN 2292-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Neferine, MedChemExpress
MedChemExpress Neferine is a major bisbenzylisoquinline alkaloid. Neferine strongly inhibits NF-κB activation.