accessibility menu, dialog, popup

UserName

CAS RN 229177-79-1

NHHNPPPhPhPhPh(?)

CAS RN 229177-79-1

IUPAC Name: N-{2'-[(diphenylphosphanyl)amino]-5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalen]-2-yl}-1,1-diphenylphosphanamine
Synonyms: N-{2'-[(diphenylphosphanyl)amino]-5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalen]-2-yl}-1,1-diphenylphosphanamine
Molecular Weight (g/mol): 660.78
Molecular Formula: C44H42N2P2
InChi Key: GTPWPBWUIWYZCB-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)C=CC(NP(C1=CC=CC=C1)C1=CC=CC=C1)=C2C1=C(NP(C2=CC=CC=C2)C2=CC=CC=C2)C=CC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results