accessibility menu, dialog, popup

UserName

CAS RN 228244-20-0

CH3H3CCH3OONN

CAS RN 228244-20-0

IUPAC Name: tert-butyl 2-cyanopyrrolidine-1-carboxylate
Synonyms: tert-butyl 2-cyanopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 196.25
Molecular Formula: C10H16N2O2
InChi Key: MDMSZBHMBCNYNO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results