CAS RN 2277-28-3
CAS RN 2277-28-3
IUPAC Name:
(2S)-2,3-dihydroxypropyl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms:
(2S)-2,3-dihydroxypropyl (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol):
354.53
Molecular Formula:
C21H38O4
InChi Key:
WECGLUPZRHILCT-GSNKCQISSA-N
SMILES:
CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC[C@@H](O)CO
1
–
1
of
1
results
1