CAS RN 2268-16-8
CAS RN 2268-16-8
IUPAC Name:
2,4,5-trifluorophenol
Synonyms:
2,4,5-trifluorophenol
Molecular Weight (g/mol):
148.08
Molecular Formula:
C6H3F3O
InChi Key:
ODGMYCITQAIRCI-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=C(F)C=C1F