CAS RN 22671-29-0
CAS RN 22671-29-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Hexanoylcarnitine, MedChemExpress
MedChemExpress L-Hexanoylcarnitine is an acylcarnitine and is found to be associated with celiac disease.