accessibility menu, dialog, popup

UserName

CAS RN 2247-88-3

NNH2F

CAS RN 2247-88-3

IUPAC Name: 3-fluoropyridin-4-amine
Synonyms: 3-fluoropyridin-4-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: UFIKBUVVVGSMGW-UHFFFAOYSA-N
SMILES: NC1=C(F)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results