CAS RN 2227425-05-8
CAS RN 2227425-05-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PF-06869206, MedChemExpress
MedChemExpress PF-06869206 is an orally bioavailable selective inhibitor of the sodium-phosphate cotransporter NaPi2a (SLC34A1) with an IC50 of 380 nM.