CAS RN 2215-21-6
IUPAC Name:
2-hydroxy-3,5-bis(propan-2-yl)benzoate
Synonyms:
2-hydroxy-3,5-diisopropylbenzoate
Molecular Weight (g/mol):
221.28
Molecular Formula:
C13H17O3
InChi Key:
XUFUYOGWFZSHGE-UHFFFAOYSA-M
SMILES:
CC(C)C1=CC(C(C)C)=C(O)C(=C1)C([O-])=O
1
–
1
of
1
results
1
3,5-Diisopropylsalicylic Acid, TRC
CAS: 2215-21-6 Molecular Formula: C13H18O3 Molecular Weight (g/mol): 222.28 Synonym: 2-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid,2-Hydroxy-3,5-bis(propan-2-yl)benzoic Acid,2-Hydroxy-3,5-diisopropylbenzoic Acid,3,5-Diisopropyl-2-hydroxybenzoic Acid IUPAC Name: 2-hydroxy-3,5-di(propan-2-yl)benzoic acid SMILES: CC(C)c1cc(C(C)C)c(O)c(c1)C(=O)O