accessibility menu, dialog, popup

UserName

CAS RN 2210-28-8

H3COOCH3CH2

CAS RN 2210-28-8

IUPAC Name: propyl 2-methylprop-2-enoate
Synonyms: propyl methacrylate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: NHARPDSAXCBDDR-UHFFFAOYSA-N
SMILES: CCCOC(=O)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results