accessibility menu, dialog, popup

UserName

CAS RN 220285-28-9

CH3OOOH

CAS RN 220285-28-9

IUPAC Name: 3-(2-ethoxyphenyl)propanoic acid
Synonyms: 3-(2-ethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: IBGXZMNEERGARC-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1CCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results