CAS RN 219727-21-6
CAS RN 219727-21-6
IUPAC Name:
ethyl 2-chloro-4-iodopyridine-3-carboxylate
Synonyms:
ethyl 2-chloro-4-iodopyridine-3-carboxylate
Molecular Weight (g/mol):
311.5
Molecular Formula:
C8H7ClINO2
InChi Key:
YOJVEUAQIMBZSL-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(I)C=CN=C1Cl
1
–
1
of
1
results
1
Ethyl 2-chloro-4-iodonicotinate, 97%, Thermo Scientific™
CAS: 219727-21-6 Molecular Formula: C8H7ClINO2 Molecular Weight (g/mol): 311.50 MDL Number: MFCD07781133 InChI Key: YOJVEUAQIMBZSL-UHFFFAOYSA-N Synonym: ethyl 2-chloro-4-iodonicotinate,3-pyridinecarboxylic acid, 2-chloro-4-iodo-, ethyl ester,2-chloro-4-iodonicotinic acid ethyl ester,2-chloro-4-iodo-nicotinic acid ethyl ester,2-chloro-4-iodopyridine-3-carboxylic acid ethyl ester,3-pyridinecarboxylicacid,2-chloro-4-iodo-,ethylester PubChem CID: 15850399 IUPAC Name: ethyl 2-chloro-4-iodopyridine-3-carboxylate SMILES: CCOC(=O)C1=C(I)C=CN=C1Cl