CAS RN 215788-65-1
CAS RN 215788-65-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II), 98%, Thermo Scientific™
CAS: 215788-65-1 Molecular Formula: C22H36Cl2FeP2Pd Molecular Weight (g/mol): 595.64 MDL Number: MFCD11656082 Synonym: [1,1'-Bis(diisopropylphosphino)ferrocene]palladium(II) chloride; PdCl2(dippf) IUPAC Name: Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II) SMILES: [Fe].Cl[Pd]Cl.CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1