accessibility menu, dialog, popup

UserName

NClClNOO

CAS RN 21427-62-3

IUPAC Name: 2,5-dichloro-3-nitropyridine
Synonyms: 2,5-dichloro-3-nitropyridine
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: OBUGJYJQJWMOQO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CN=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)