CAS RN 21416-43-3
CAS RN 21416-43-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Willardiine, MedChemExpress
MedChemExpress (S)-Willardiine is a potent agonist of AMPA/kainate receptors with EC50 of 44.8 uM.