accessibility menu, dialog, popup

UserName

OOHN

CAS RN 2124-55-2

IUPAC Name: 1H-indole-4-carboxylate
Synonyms: 1H-indole-4-carboxylate
Molecular Weight (g/mol): 160.15
Molecular Formula: C9H6NO2
InChi Key: ROGHUJUFCRFUSO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C2C=CNC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)