accessibility menu, dialog, popup

UserName

CAS RN 21239-29-2

H3COOCH3H3C

CAS RN 21239-29-2

IUPAC Name: ethyl 3,5-dimethylbenzoate
Synonyms: ethyl 3,5-dimethylbenzoate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: IHAAVLXHNOZMBC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)