accessibility menu, dialog, popup

UserName

FFFOFFF

CAS RN 21084-22-0

IUPAC Name: [3-(trifluoromethyl)phenyl][4-(trifluoromethyl)phenyl]methanone
Synonyms: [3-(trifluoromethyl)phenyl][4-(trifluoromethyl)phenyl]methanone
Molecular Weight (g/mol): 318.22
Molecular Formula: C15H8F6O
InChi Key: HWBXNAPXNCBCQO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results